Skip to Content
Merck

Skip To

SK009

AROCLOR 1268 CAL STANDARD

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200


grade

certified reference material

Quality Segment

format

single component solution

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.


Analyte

Description

  • Aroclor-1268 (PCB-1268) 0.1 µg/mL

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

10.4 °F - closed cup

flash_point_c

-11.99 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library