Merck
Metabolite Libraries
Close-up photograph of multi-pipettes adding samples to the deep well tray of a metabolite library kit with a violet solution in a laboratory.
METABOLITE LIBRARIES AND MIXTURES
Metabolite libraries and mixtures provide a convenient solution for identifying hundreds of metabolites in metabolomics testing and analysis, primarily for metabolite qualification and quantification in MS and NMR, functional cellular assays, and phenotypic screening. Our portfolio includes:
  • Ready-to-use standard mixture solutions for various research purposes, such as quantification, disease-related metabolite studies, or quality control samples in LC-MS.
  • IROA Technologies plated metabolite libraries containing numerous diverse classes of metabolites, each individually placed in a 96-well plate. Each library includes a user manual, plate map, and MLSDiscoveryTM software for data extraction, manipulation, and storage for building internal libraries.
  • Small molecule metabolite libraries and kits covering key metabolic pathways, including a Glycolysis/Gluconeogenesis kit, Pentose Phosphate kit, TCA Cycle kit, and Urea Cycle kit.
Read more about
Products
Sort by Relevance
Showing 1-20 of 46
Change View
Showing 1-20 of 46
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 46 results
METABOLITE LIBRARY OF STANDARDS
Our collection of metabolite libraries, supplied by IROA Technologies, contains a broad range of metabolite classes, each individually placed in a 96-well plate. These high purity (>95%) compounds are conveniently supplied in seven polypropylene racks of 96-well plates, with each well containing 5μg dried weight. The metabolites can be used as standards and injected individually or pooled, allowing for multiple mixtures to be analyzed per injection. 
Individual plate compounds can also be ordered separately to fulfill your research needs better.
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sorry, an unexpected error has occurred
Network error: Failed to fetch
TRUQUANT Yeast Extract Workflow Kit
TruQuant Yeast Extract Workflow Kit is a sophisticated quantitation system designed to take simultaneous and accurate biological measurements of several hundred biochemicals in small biological samples. The TruQuant workflow kit combines a software solution with a comprehensive set of over 500 complex internal standards to address issues related to ionization inefficiencies and matrix effects. This approach enables suppression-corrected data, ensures consistent quantitative results across different platforms, facilitates validated compound identification, and enables ongoing quality assurance/quality control of instruments.
Related Resources
  • Protocol: Mass Spectrometry Metabolite Library
    MSMLS™ offers high-purity small biochemical molecules for broad primary metabolism studies in an economical, ready-to-use format.
  • Brochure: Mass Spectrometry Metabolite Library
    MSMLS (Mass Spectrometry Metabolite Library) is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (95%) compounds supplied in an economical, ready-to-use format.
  • Brochure: IROA TruQuant IQQ Workflow Kit
    The IROA TruQuant IQQ Workflow Kit is a high-quality quantitation system for making simultaneous accurate biological measurements on several hundred biochemicals in small quantities of biological samples.
  • Brochure: Metabolomics
    Integrated metabolomics workflows provide efficient compound profiling, pinpointing, and designation for targeted or untargeted studies.