Skip to Content
Merck
All Photos(1)

Key Documents

M9802

Millipore

Malt agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CHF 210.00

CHF 210.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
CHF 210.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

CHF 210.00


Estimated to ship on30 May 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.00 g/L
Malt extract, 30.00 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

5.5±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
food and beverages

microbiology

suitability

selective by low pH for Aspergillus spp.
selective by low pH for Candida spp.
selective by low pH for Penicillium spp.
selective by low pH for Pichia spp.
selective by low pH for Saccharomyces spp.
selective by low pH for Zygosaccharomyces spp.

Application

Used for the detection and isolation of yeasts and molds from dairy products and foods.

Preparation Note

Suspend 45 g in 1000 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Avoid overheating as it will result in a softer and darker agar.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Baietto et al.
Sensors (Basel, Switzerland), 10(2), 1062-1092 (2010-01-01)
Tree assessment methodologies, currently used to evaluate the structural stability of individual urban trees, usually involve a visual analysis followed by measurements of the internal soundness of wood using various instruments that are often invasive, expensive, or inadequate for use

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service