Skip to Content
Merck
All Photos(1)

Key Documents

Y3752

Millipore

Yeast malt broth

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CHF 298.00

CHF 298.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
CHF 298.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

CHF 298.00


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

composition

Dextrose, 10.00 g/L
Malt extract, 3.00 g/L
Peptic digest of animal tissue, 5.00 g/L
Yeast extract, 3.00 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.2±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
environmental
food and beverages

microbiology

suitability

nonselective for molds (General Media)
nonselective for yeasts (General Media)

Application

Used for the isolation and cultivation of yeasts and molds.

Preparation Note

Suspend 21 g of Yeast Malt Broth in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. For preparing a selective media acidify the media to pH 3.0-4.0, or add the appropriate antibiotic. Do not heat the media after addition of acid or antibiotics.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rucha Natu et al.
Analytical chemistry, 91(7), 4357-4367 (2019-03-05)
Cell sorting methods are required in numerous healthcare assays. Although flow cytometry and magnetically actuated sorting are widespread techniques for cell sorting, there is intense research on label-free techniques to reduce the cost and complexity of the process. Among label-free
Rucha Natu et al.
Lab on a chip, 19(15), 2512-2525 (2019-07-02)
Selective manipulation of single cells is an important step in sample preparation for biological analysis. A highly specific and automated device is desired for such an operation. An ideal device would be able to selectively pick several single cells in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service