Skip to Content
Merck
All Photos(3)

Key Documents

920002

Sigma-Aldrich

CX85

Umicore

Synonym(s):

Dichlorobis[(tert-butyldiphenyl)phosphine]palladium(II), Pd(P(tBu)Ph2)2Cl2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 100.00

CHF 100.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
CHF 100.00

About This Item

Empirical Formula (Hill Notation):
C32H38Cl2P2Pd
CAS Number:
Molecular Weight:
661.92
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

CHF 100.00


Estimated to ship on30 April 2025


Request a Bulk Order

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

227-228 °C

SMILES string

[Pd]([PH](C(C)(C)C)(c4ccccc4)c3ccccc3)([PH](C(C)(C)C)(c2ccccc2)c1ccccc1)(Cl)Cl

InChI

1S/2C16H19P.2ClH.Pd/c2*1-16(2,3)17(14-10-6-4-7-11-14)15-12-8-5-9-13-15;;;/h2*4-13H,1-3H3;2*1H;

InChI key

QZOYEMHPCYSVBE-UHFFFAOYSA-N

Application

Umicore CX85 is a homogeneous catalyst useful for cross-coupling reactions with arenes, especially for Suzuki-Miyaura cross-couplings.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service