Skip to Content
Merck
All Photos(1)

Key Documents

90007

Supelco

Glyphosate-2-13C

PESTANAL®, analytical standard

Synonym(s):

N-(Phosphonomethyl)glycine-2-13C

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 192.00

CHF 192.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 192.00

About This Item

Linear Formula:
HO2C13CH2NHCH2PO(OH)2
CAS Number:
Molecular Weight:
170.07
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

CHF 192.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+1

SMILES string

OC(=O)[13CH2]NCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1

InChI key

XDDAORKBJWWYJS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

13C enrichment ≥98.5 atom% based upon labeld precursor

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Glyphosate analysis: LC-MS/MS method with Supel™ Carbon LC U/HPLC column for stability and retention.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service