Skip to Content
Merck
Glyphosate-2-13C PESTANAL®, analytical standard

Glyphosate-2-13C

Glyphosate-2-13C PESTANAL®, analytical standard
Synonym(s):
N-(Phosphonomethyl)glycine-2-13C
Linear Formula:
HO2C13CH2NHCH2PO(OH)2
CAS Number:
Molecular Weight:
170.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing