Skip to Content
Merck
All Photos(1)

Key Documents

74867

Supelco

Dichlorvos-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

2,2-Dichlorovinyl di(methyl-d3) phosphate, Dichlorvos-d6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 359.00

CHF 359.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 359.00

About This Item

Empirical Formula (Hill Notation):
C4D6HCl2O4P
CAS Number:
Molecular Weight:
227.01
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

CHF 359.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥95.0 atom % D

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

ClC(Cl)=COP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=O

InChI

1S/C4H7Cl2O4P/c1-8-11(7,9-2)10-3-4(5)6/h3H,1-2H3/i1D3,2D3

InChI key

OEBRKCOSUFCWJD-WFGJKAKNSA-N

General description

Dichlorvos-(dimethyl-d6) is an isotope labeled analog of the organophosphate insecticide dichlorvos, wherein the dimethoxy protons are replaced by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semiconductor-sensitized photodegradation of dichlorvos in water using TiO2 and ZnO as catalysts
Evgenidou E, et al.
Applied Catalysis. B, Environmental, 59(1-2), 81-89 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service