Skip to Content
Merck
All Photos(1)

Documents

94093

Supelco

Parathion-methyl-d6 (dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

O,O-Dimethyl-d6 O-(4-nitrophenyl) phosphorothioate, Methyl parathion-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D6H4NO5PS
CAS Number:
Molecular Weight:
269.24
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥99 atom % D

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas (Argon)

application(s)

agriculture
environmental

format

neat

SMILES string

S=P(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])OC1=CC=C([N+]([O-])=O)C=C1

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3/i1D3,2D3

InChI key

RLBIQVVOMOPOHC-WFGJKAKNSA-N

General description

Parathion-methyl-d6 (dimethyl-d6) is an isotope-labeled analog of the organophosphorus insecticide, parathion-methyl.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Analysis Note

identification (LC-MS): identity conforms

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - open cup

Flash Point(C)

46 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fluorescence polarization immunoassay based on a monoclonal antibody for the detection of the organophosphorus pesticide parathion-methyl
Kolosova AY, et al.
Journal of Agricultural and Food Chemistry, 51(5), 1107-1114 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service