Skip to Content
Merck
All Photos(1)

Key Documents

63655

Supelco

(+)-p-Menth-1-ene

analytical standard

Synonym(s):

(R)-(+)-4-Isopropyl-1-methylcyclohexene, Carvomenthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
3194398
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +110±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.457

bp

175-177 °C (lit.)

density

0.823 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC(C)[C@@H]1CCC(C)=CC1

InChI

1S/C10H18/c1-8(2)10-6-4-9(3)5-7-10/h4,8,10H,5-7H2,1-3H3/t10-/m0/s1

InChI key

FAMJUFMHYAFYNU-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Used as starting material in EPC syntheses; ring cleavage at the double bond by ozonolysis; Or via epoxidation; Trans-selective ene-reaction with acrylates

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Negin Farshchi et al.
Journal of chromatographic science, 56(8), 671-678 (2018-05-12)
Limonene is a colorless liquid hydrocarbon and had been investigated as a plasticizer for many plastics. Prediction of solubility between different materials is an advantage in many ways, one of the most convenient ways to know the compatibility of materials
F. Nicotra et al.
The Journal of Organic Chemistry, 51, 1272-1272 (1986)
J.V. Duncia et al.
Journal of the American Chemical Society, 104, 1930-1930 (1982)
T. Kitahara et al.
Tetrahedron, 43, 2689-2689 (1987)
J.D. White et al.
Journal of the American Chemical Society, 103, 1813-1813 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service