Skip to Content
Merck
All Photos(1)

Key Documents

ERMEB530C

Al alloy

0.1% Au, 1.0 mm wire, ERM®, certified reference material

Synonym(s):

Aluminum alloy

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 434.00

CHF 434.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
CHF 434.00

About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 434.00


Available to ship on22 April 2025Details


Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

matrix material

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified for the mass fraction of Au.

Analysis Note

For more information please see:
ERMEB530C

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service