Skip to Content
Merck
All Photos(2)

Documents

32983

Supelco

Enrofloxacin-d5 hydrochloride

VETRANAL®, analytical standard

Synonym(s):

1-Cyclopropyl-7-(4-ethyl-d5-1-piperazinyl)-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D5H17FN3O3 · HCl
Molecular Weight:
400.89
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])N1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

Application

Enrofloxacin-d5 hydrochloride may be used as a reference standard in the determination of enrofloxacin in dairy products using ultra high performance liquid chromatography coupled with tandem mass spectrometry (UHPLC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development and Validation of a Multi-class UHPLC-MS/MS Method for Determination of Antibiotic Residues in Dairy Products.
Schwaiger B, et al.
Food Analytical Methods, 1-18 (2018)
Đorđe Tadić et al.
Analytical and bioanalytical chemistry, 411(20), 5209-5222 (2019-06-12)
The developed method was evaluated for the determination of 10 antibiotics belonging to four chemical classes (fluoroquinolones, sulfonamides, lincosamides, and metoxybenzylpyrimidines) and six of their metabolites in four vegetable matrices (lettuce, tomato, cauliflower, and broad beans). The reported method detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service