Skip to Content
Merck
All Photos(1)

Documents

05939

Millipore

Rabbit Plasma Fibrinogen Supplement

suitable for microbiology

Synonym(s):

RPF Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

Agency

according to ISO 6888-2:2020

Quality Level

sterility

sterile (Filtered and Aseptic Handled)

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

General description

Rabbit Plasma Fibrinogen Supplement is used for isolation, identification, confirmation, and enumeration of coagulase positive Staphylococcus aureus from food and other specimens. It is used with Baird-Parker Agar Base. The reduction of potassium tellurite concentration in RPF supplement results in S. aureus forming grey or black colonies, surrounded by an opaque halo of precipitation, i.e. the coagulase reaction.

Application

Rabbit Plasma Fibrinogen Supplement is used for isolation, identification, confirmation, and enumeration of coagulase positive Staphylococcus aureus from food and other specimens.

Components

(per vial, sufficient for 100 ml medium)
Rabbit plasma 2.5ml
Bovine Fibrinogen 0.5g
Trypsin inhibitor 2.5mg
Potassium tellurite 2.5mg

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service