Skip to Content
Merck
All Photos(1)

Key Documents

30-0770

Sigma-Aldrich

Tetraethylammonium bromide

SAJ special grade, ≥98.0%

Synonym(s):

TEA bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
Beilstein:
3563430
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
grade:
SAJ special grade

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

pH

5.0-7.0 (25 °C, 100 g/L)

mp

285 °C (dec.) (lit.)

SMILES string

[Br-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

361.4 °F - closed cup

Flash Point(C)

183 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Mruk et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(34), 13579-13583 (2012-08-08)
Calmodulin (CaM) is a ubiquitous intracellular calcium sensor that directly binds to and modulates a wide variety of ion channels. Despite the large repository of high-resolution structures of CaM bound to peptide fragments derived from ion channels, there is no
Min Shen et al.
Vascular pharmacology, 58(1-2), 64-70 (2012-07-24)
Evidence has indicated that resveratrol (Res) produces vasorelaxation and may decrease the coronary heart disease mortality. However, several pathways involved in the mechanism of vasorelaxation are still unclear. This study was designed, therefore, to test the probable ion channels or
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Jayabalan Nirmal et al.
Experimental eye research, 116, 27-35 (2013-07-31)
The present study was conducted to test the hypothesis; OCT may be active from blood-to-vitreous for the uptake of its substrates. Ocular uptake of Tetraethylammonium (TEA) across blood ocular barriers and the tissue distribution was evaluated in vivo in New
Lei Yang et al.
The Journal of general physiology, 140(5), 529-540 (2012-10-31)
We investigated the effects of changing extracellular K(+) concentrations on block of the weak inward-rectifier K(+) channel Kir1.1b (ROMK2) by the three intracellular cations Mg(2+), Na(+), and TEA(+). Single-channel currents were monitored in inside-out patches made from Xenopus laevis oocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service