Merck
Structural insights into neuronal K+ channel-calmodulin complexes.
Proceedings of the National Academy of Sciences of the United States of America (2012-08-08)
Karen Mruk, Shiven M D Shandilya, Robert O Blaustein, Celia A Schiffer, William R Kobertz
ABSTRACT
Calmodulin (CaM) is a ubiquitous intracellular calcium sensor that directly binds to and modulates a wide variety of ion channels. Despite the large repository of high-resolution structures of CaM bound to peptide fragments derived from ion channels, there is no structural information about CaM bound to a fully folded ion channel at the plasma membrane. To determine the location of CaM docked to a functioning KCNQ K(+) channel, we developed an intracellular tethered blocker approach to measure distances between CaM residues and the ion-conducting pathway. Combining these distance restraints with structural bioinformatics, we generated an archetypal quaternary structural model of an ion channel-CaM complex in the open state. These models place CaM close to the cytoplasmic gate, where it is well positioned to modulate channel function.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)