Skip to Content
Merck
All Photos(1)

Key Documents

28-3440

Sigma-Aldrich

Sodium nitrate

JIS special grade, ≥99.0%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
solid
solubility:
water: soluble 874 g/L at 20 °C
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

5.5-8.0 (20 °C, 50 g/L)

mp

306 °C (dec.) (lit.)

solubility

water: soluble 874 g/L at 20 °C

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Daryoush Afzali et al.
Journal of chromatography. A, 1247, 35-41 (2012-06-08)
In the present study, a new approach which uses immunoaffinity column clean-up combined with dispersive liquid-liquid microextraction (DLLME) is proposed for the preconcentration of ultra trace amounts of aflatoxins (B₁, B₂, G₁ and G₂). The aflatoxins are then determined using
Raúl Bescós et al.
Medicine and science in sports and exercise, 44(12), 2400-2409 (2012-07-20)
Supplementation with inorganic nitrate has been suggested to be an ergogenic aid for athletes as nitric oxide donor. The purpose of this study was to determine whether ingestion of inorganic sodium nitrate benefits well-trained athletes performing a 40-min exercise test
Ibrahim Yazji et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9451-9456 (2013-05-08)
Necrotizing enterocolitis (NEC) is a devastating disease of premature infants characterized by severe intestinal necrosis and for which breast milk represents the most effective protective strategy. Previous studies have revealed a critical role for the lipopolysaccharide receptor toll-like receptor 4
Shao-Ju Chien et al.
Translational research : the journal of laboratory and clinical medicine, 163(1), 43-52 (2013-10-12)
Nitric oxide (NO) deficiency mediates oxidative stress in the kidney and is involved in the development of hypertension. NO synthesis occurs via 2 pathways: nitric oxide synthase (NOS) dependent and NOS-independent. We tested whether the development of hypertension is prevented

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service