01-4430
Ammonium bicarbonate
SAJ first grade
Synonym(s):
Ammonium hydrogen carbonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
SAJ first grade
vapor density
2.7 (vs air)
vapor pressure
513 hPa ( 50 °C)
67.1 hPa ( 20 °C)
form
solid
availability
available only in Japan
pH
7.0-8.5 (25 °C, 79.1 g/L)
storage temp.
15-25°C
SMILES string
N.OC(O)=O
InChI
1S/CH2O3.H3N/c2-1(3)4;/h(H2,2,3,4);1H3
InChI key
ATRRKUHOCOJYRX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Toxins, 7(3), 936-950 (2015-03-21)
Cnidarian venom research has lagged behind other toxinological fields due to technical difficulties in recovery of the complex venom from the microscopic nematocysts. Here we report a newly developed rapid, repeatable and cost effective technique of venom preparation, using ethanol
Chemical research in toxicology, 22(10), 1713-1720 (2009-08-21)
The arsenic (+3 oxidation state) methyltransferase (As3mt) gene encodes a 43 kDa protein that catalyzes methylation of inorganic arsenic. Altered expression of AS3MT in cultured human cells controls arsenic methylation phenotypes, suggesting a critical role in arsenic metabolism. Because methylated
PloS one, 9(4), e95007-e95007 (2014-04-17)
Protozoan parasites of Leishmania genus are able to successfully infect their host macrophage due to multiple virulence strategies that result in its deactivation. Recent studies suggest Leishmania GP63 to be a critical virulence factor in modulation of many macrophage molecules
Journal of controlled release : official journal of the Controlled Release Society, 152(3), 370-375 (2011-03-15)
Spray-dried powders for lung delivery of sodium alendronate (SA) were prepared from hydroalcoholic solutions. Formulations display geometric particle size below to 12 μm and spherical shape associated to a hollow structure. The addition of leucine and ammonium bicarbonate leads to
EMBO molecular medicine, 7(9), 1166-1178 (2015-08-09)
Non-invasive detection of colorectal cancer with blood-based markers is a critical clinical need. Here we describe a phased mass spectrometry-based approach for the discovery, screening, and validation of circulating protein biomarkers with diagnostic value. Initially, we profiled human primary tumor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service