Skip to Content
Merck
All Photos(1)

Key Documents

11667475001

Roche

Anti-HA-Peroxidase

from mouse IgG2bκ

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12CA5, monoclonal

Assay

≥95% (HPLC and SDS-PAGE)

form

solution

packaging

pkg of 50 μg (500 μl)

manufacturer/tradename

Roche

concentration

0.1 mg/mL

isotype

IgG2bκ

epitope sequence

YPYDVPDYA

storage temp.

−20°C

General description

Anti-HA-Peroxidase is a monoclonal antibody to the HA peptide, conjugated to peroxidase. Anti-HA-Peroxidase recognizes the HA nonapeptide sequence (YPYDVPDYA), derived from the human influenza virus hemagglutinin protein (amino acids 98-106). The antibody recognizes its antigenic determinant even when the HA peptide epitope is introduced into unrelated recombinant proteins by a technique known as "epitope tagging".

Specificity

Anti-HA-Peroxidase recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins regardless of its position (N-terminal, C-terminal or internal).
The antibody reacts with the HA epitope, a nonapeptide sequence (YPYDVPDYA) derived from the influenza hemagglutinin protein.

Immunogen

Amino acids 98-106 from the human influenza virus hemagglutinin protein

Application

  • Use Anti-HA-Peroxidase for the immunochemical detection of native influenza hemagglutinin protein and recombinant HA-tagged proteins using:
  • Dot blot
  • Western blot
Use of Anti-HA-Peroxidase eliminates the need for a secondary detection step.
Note: For experiments in which sensitivity is not critical, use Anti-HA-Peroxidase (12CA5). For higher sensitivity detection in western blotting at 10-fold lower concentration, use Anti-HA High Affinity.

Quality

Each lot of Anti-HA antibody is tested for functionality and purity relative to a reference standard to confirm the quality of each new reagent preparation.

Preparation Note

Stabilizers: BSA, sucrose, 0.02% thymol
Working concentration: Working concentration of conjugate depends on application and substrate.


The following concentrations should be taken as a guideline:
  • Western blot: 0.1 μg/ml

Working solution: 20-fold dilution of Western Blocking Reagent in 1x PBS.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asvin K K Lakkaraju et al.
Molecular biology of the cell, 23(14), 2712-2722 (2012-06-01)
Mammalian cells secrete a large number of small proteins, but their mode of translocation into the endoplasmic reticulum is not fully understood. Cotranslational translocation was expected to be inefficient due to the small time window for signal sequence recognition by
Sonia V del Rincón et al.
Oncogene, 23(57), 9269-9279 (2004-11-02)
Insulin receptor substrate-1 (IRS-1) mediates signaling from the insulin-like growth factor type-I receptor. We found that all-trans retinoic acid (RA) decreases IRS-1 protein levels in MCF-7, T47-D, and ZR75.1 breast cancer cells, which are growth arrested by RA, but not
Marta García-León et al.
The Plant cell, 31(10), 2411-2429 (2019-08-01)
The plant endosomal trafficking pathway controls the abundance of membrane-associated soluble proteins, as shown for abscisic acid (ABA) receptors of the PYRABACTIN RESISTANCE1/PYR1-LIKE/REGULATORY COMPONENTS OF ABA RECEPTORS (PYR/PYL/RCAR) family. ABA receptor targeting for vacuolar degradation occurs through the late endosome
Ryan M Drenan et al.
The Journal of biological chemistry, 281(38), 28222-28231 (2006-07-27)
The RGS7 (R7) family of G protein regulators, Gbeta5, and R7BP form heterotrimeric complexes that potently regulate the kinetics of G protein-coupled receptor signaling. Reversible palmitoylation of R7BP regulates plasma membrane/nuclear shuttling of R7*Gbeta5*R7BP heterotrimers. Here we have investigated mechanisms
Helen Wedegaertner et al.
Molecular biology of the cell, 34(9), ar93-ar93 (2023-05-24)
The α-arrestin ARRDC3 is a recently discovered tumor suppressor in invasive breast cancer that functions as a multifaceted adaptor protein to control protein trafficking and cellular signaling. However, the molecular mechanisms that control ARRDC3 function are unknown. Other arrestins are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service