Skip to Content
Merck
All Photos(2)

Key Documents

MABN665

Sigma-Aldrich

Anti-Synaptotagmin-7 Antibody, clone N275/14

clone N275/14, from mouse

Synonym(s):

Synaptotagmin-7, Synaptotagmin VII, SytVII

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 424.00

CHF 424.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 424.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 424.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N275/14, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYT7(9066)

General description

Synaptotagmin-7 is also referred to as Synaptotagmin VII (SytVII) and is a member of the synaptotagmin family of transmembrane transporters. Synaptotagmin-7 is involved in plasma membrane repair, and possibly Ca2+-dependent exocytosis and traffiking of secretory vesicles.

Specificity

This antibody recognizes the cytoplasmic C2A domain of Synaptotagmin-7. This antibody does not cross react with Synaptotagmin-6 or other Synaptotagmins (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Epitope: Cytoplasmic C2A domain
Recombinant protein corresponding to the cytoplasmic C2A domain of mouse Synaptotagmin-7.

Application

Anti-Synaptotagmin-7 Antibody, clone N275/14 is a highly specific mouse monoclonal antibody, that targets Synaptotagmin & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Synaptotagmin-7 in human thalamus tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Synaptotagmin-7 in 10 µg of mouse brain tissue lysate.

Target description

~ 45 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synaptotagmin-7 Enhances Facilitation of Cav2.1 Calcium Channels.
Djillani, et al.
eNeuro, 9 (2023)
Gaël Barthet et al.
Nature communications, 9(1), 4780-4780 (2018-11-16)
Mutations of the intramembrane protease presenilin (PS) or of its main substrate, the amyloid precursor protein (APP), cause early-onset form of Alzheimer disease. PS and APP interact with proteins of the neurotransmitter release machinery without identified functional consequences. Here we
Christopher Weyrer et al.
Cell reports, 36(12), 109719-109719 (2021-09-23)
Synaptotagmin 7 (Syt7) is a high-affinity calcium sensor that is implicated in multiple aspects of synaptic transmission. Here, we study the influence of Syt7 on the climbing fiber (CF) to Purkinje cell (PC) synapse. We find that small facilitation and
Bassam Tawfik et al.
eLife, 10 (2021-03-23)
Synaptotagmins confer calcium-dependence to the exocytosis of secretory vesicles, but how coexpressed synaptotagmins interact remains unclear. We find that synaptotagmin-1 and synaptotagmin-7 when present alone act as standalone fast and slow Ca2+-sensors for vesicle fusion in mouse chromaffin cells. When
Katarina Trajkovic et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(37), 9000-9012 (2017-08-20)
Huntington's disease (HD) is an autosomal-dominant neurodegenerative disorder caused by the expansion of a CAG triplet in the gene encoding for huntingtin (Htt). The resulting mutant protein (mHtt) with extended polyglutamine (polyQ) sequence at the N terminus leads to neuronal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service