Skip to Content
Merck
All Photos(1)

Key Documents

MABF2108

Sigma-Aldrich

Anti-Chlamydial HSP60 Antibody, clone A57-B9

clone A57-B9, from mouse

Synonym(s):

Chlamydia heat shock protein 60

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 400.00

CHF 400.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 400.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 400.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

A57-B9, monoclonal

species reactivity

bacteria, Chlamydia

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

target post-translational modification

unmodified

General description

Chlamydia trachomatis is a Gram-negative bacterium that is responsible to sexually transmitted diseases leading to pelvic inflammatory disease, ectopic pregnancy, infertility, and outbreaks of trachoma-associated blindness and lymphogranuloma venereum (LGV). Chlamydia trachomatis consists of eighteen different serological variants (serovars) that include a few subvariants. These are identified based on serological reactivity of the epitopes on their outer membrane. Intracellularly chlamydia replicates within a vacuole. Chlamydia infection is initiated with the expression of a chlamydial early gene product(s), which isolate the inclusion from the endocytic-lysosomal pathway and makes it fusogenic with sphingomyelin-containing exocytic vesicles. This change in vesicular interaction allows the delivery of the vacuole to the peri-Golgi region of the host cell. Antigens from all members of the Chlamydia genus display heat resistance and sensitivity to oxidation by sodium periodate. Clone A57-B9 detects Chlamydial HSP60 (GroEL). It reacts with the short peptide of 6 amino acids from the carboxyl terminal region of HSP60. HSP60 facilitates the correct folding of imported proteins and may prevent misfolding and promote the refolding and proper assembly of unfolded polypeptides generated under stress conditions.

Specificity

Clone A57-B9 detects Chlamydial HSP60. It targets an epitope with in 144 amino acids in the carboxy terminal of HSP60.

Immunogen

Recombinant Chlamydia trachomatis serovar A HSP60 emulsified in complete Freund adjuvant.

Application

Anti-Chlamydial HSP60, clone A57-B9, Cat. No. MABF2108, is a mouse monoclonal antibody that detects Chlamydial HSP60 and has been tested for use in ELISA, Immunofluorescence, Immunocytochemistry, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected Chlamydial HSP60 in Immunocytochemistry applications (Yuan, Y., et. al. (1992). Infect Immun. 60(6):2288-96).

Immunofluorescence Analysis: A representative lot detected Chlamydial HSP60 in Immunofluorescence applications (Southern, T., et. al. (2012). Clin Vaccine Immunol. 19(11):1864-9).

ELISA Analysis: A representative lot detected Chlamydial HSP60 in ELISA applications (Morrison, S.G., et. al. (2005). J Immunol. 175(11):7536-42).

Western Blotting Analysis: A representative lot detected chlamydial HSP60 in Western Blotting applications (Hechard, C., et. al. (2004). J Med Microbiol. 53(Pt 9):861-8; LaVerda, D., et. al. (1997). J Clin Microbiol. 35(5):1209-15; Southern, T., et. al. (2012). Clin Vaccine Immunol. 19(11):1864-9; Yuan, Y., et. al. (1992). Infect Immun. 60(6):2288-96).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in lysates from HeLa cells infected with chlamydia trachomatis serovar L2 (LGV 434 L2) .

Western Blotting Analysis: 2 µg/mL of this antibody detected Chlamydial HSP60 in lysates from HeLa cells infected with Chlamydia trachomatis serovar L2 (LGV 434 L2) .

Target description

~60 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service