Skip to Content
Merck
All Photos(1)

Key Documents

Z374423

Sigma-Aldrich

Scienceware® electrophoresis plate rack, coated steel

7 slots for single plates and 3 slots for double plates, pkg of 1 ea

Synonym(s):

electrophoresis plate storage

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 151.00

CHF 151.00


Check Cart for Availability


Select a Size

Change View
1 EA
CHF 151.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

CHF 151.00


Check Cart for Availability

material

epoxy (coated steel)

grade

electrophoresis grade

feature

Holds 3 x double plates
Holds 7 x plates

packaging

pkg of 1 ea

manufacturer/tradename

Bel-Art F13595-0000

L × W × H

20 cm (7.75 in.) × 16 cm (6.25 in.) × 18 cm (7.5 in.)

General description

Offers unmatched versatility for conveniently drying and storing up to 10 electrophoresis plates (7 slots for single plates and 3 slots for double plates). Made of epoxy-coated heavy-duty steel, with rubber pads on the bottom of the rack to keep the rack from sliding.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service