Skip to Content
Merck
All Photos(2)

Key Documents

FCMAB110P

Sigma-Aldrich

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12 PE conjugate

clone 10H11.E12, from mouse, PE

Synonym(s):

A-T, mutated

AT mutated

TEL1, telomere maintenance 1, homolog

ataxia telangiectasia mutated

ataxia telangiectasia mutated (includes complementation groups A, C and D)

ataxia telangiectasia mutated protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
CHF 309.00

CHF 309.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
CHF 309.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 309.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10H11.E12, monoclonal

species reactivity

rat, mouse, human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer1981)

Gene Information

human ... ATM(472)

Related Categories

General description

N-terminal c-Myc, GST-tagged, recombinant human Cardiac Troponin I full length, expressed by baculovirus in Sf21 insect cells. Purified using glutathione sepharose.

Specificity

Antibody recognizes ATM phosphorylated at Ser1981.
Predicted to cross-react with rat based on sequence homology

Immunogen

Epitope: Phosphorylated at and around Ser1981
KLH-conjugated, synthetic peptide corresponding to human ATM phosphorylated at Ser1981.

Application

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12 PE conjugate detects level of phospho-ATM (Ser1981) & has been published & validated for use in FC.

Quality

Evaluated by Flow Cytometry with HeLa cells.

Target description

~370 kDa Calculated

Physical form

Purified mouse monoclonal IgG1κ conjugated to phycoerythrin in PBS with less than 0.09% sodium azide and 15 mg/mL BSA.

Analysis Note

Control
Irradiated HeLa cells

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiting Lim et al.
International journal of radiation oncology, biology, physics, 84(3), 800-806 (2012-03-27)
We have previously shown that the antimalarial agent chloroquine can abrogate the lethal cellular effects of low-dose-rate (LDR) radiation in vitro, most likely by activating the ataxia-telangiectasia mutated (ATM) protein. Here, we demonstrate that chloroquine treatment also protects against lethal doses
S Masneri et al.
Stem cell research, 41, 101596-101596 (2019-11-02)
Using a Sendai Virus based vector delivering Yamanaka Factors, we generated induced Pluripotent Stem Cells (iPSCs) from peripheral blood mononuclear cells of a patient affected by Ataxia Telangiectasia (AT), caused by a novel homozygous deletion in ATM, spanning exons 5-7.
Sabrina Fritah et al.
Cancers, 12(9) (2020-09-16)
Resistance to chemotherapy by temozolomide (TMZ) is a major cause of glioblastoma (GBM) recurrence. So far, attempts to characterize factors that contribute to TMZ sensitivity have largely focused on protein-coding genes, and failed to provide effective therapeutic targets. Long noncoding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service