Skip to Content
Merck
All Photos(4)

Key Documents

EZFITMHTA3

Millipore

EZ-Fit® Manifold Filtration Head

for use with EZ-Fit® Filtration Manifold, For use with EZ-Fit® filtration units, 3 units, pkg of 3 unit

Synonym(s):

EZ-Fit® Manifold Filtration Head, Filtration Head, For bioburden application, filtration head for manifold, Manifold Head

Sign Into View Organizational & Contract Pricing

Select a Size

3 UNITS
CHF 837.00

CHF 837.00


Check Cart for Availability

Request more information

Select a Size

Change View
3 UNITS
CHF 837.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

CHF 837.00


Check Cart for Availability

Request more information

material

316L stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EZ-Fit® Manifold Filtration Head is an integral part of EZ-Fit® filtration system designed to hold the EZ-Fit® Funnels during filtration process. The manifold head is autoclavable and easy to de-assemble thereby limiting chances of any biofilm formation. The silicone O-ring enables proper fitting of the head to the EZ-Fit® Manifold system.

Application

EZ-Fit® Manifold Filtration Head is used to hold the EZ-Fit® Funnels during routine filtration process. Liquid samples of raw materials, in process and finished products from diverse industries can be tested using the equipment.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service