Skip to Content
Merck
All Photos(1)

Documents

EZFITVALV1

Millipore

EZ-Fit® Manifold complete valve

EZ-Fit® Manifold accessory, suitable (general microbial analysis)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

input

sample type beer
sample type cosmetics
sample type pharmaceutical(s)
sample type bottled water

suitability

suitable (general microbial analysis)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Fit® Filtration Unit

shipped in

ambient

Related Categories

General description

EZ-Fit® Manifold complete valve is integrated into the flexible tubing connection to avoid liquid back-flow from the vacuum system. The 2-ways valves are large and easy to operate. The design of the 2-way valve prevents the user accidentally touching the filtration head while opening or closing the valve. Filtration heads, valves, and tubing connections are all easily removable and can be easily cleaned.

Application

EZ-Fit® Manifold complete valve prevents the liquid back-flow from the vacuum system in the EZ-Fit® Manifold.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service