Skip to Content
Merck
All Photos(3)

Key Documents

ABN180

Sigma-Aldrich

Anti-TFG Antibody

from rabbit

Synonym(s):

Protein TFG, TRK-fused gene protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 351.00

CHF 351.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 351.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 351.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TFG(10342)

General description

The TRK-fused gene protein (TFG) gene is highly conserved among mammalian species, and is widely studied as a fusion gene partnering with various oncogenes. The function of the resultant TFG protein product is still unclear. TFG regulates the activity of phosphotyrosine-specific phosphatase-1 (SHP-1), and may be important in cells containing SHP-1 such as epithelial and neuronal cells. Defective TFG contributes to thyroid papillary carcinoma.

Specificity

This antibody recognizes the C-terminus of TFG.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human TFG.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-TFG Antibody is validated for use in Western Blotting, IHC(P) for the detection of TFG.
Western Blot Analysis: 0.1 µg/mL from a representative lot detected TFG on 10 µg of human brain tissue lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected TFG in mouse hippocampus, mouse cerebellum, human placenta, and human pancreas tissues.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected TFG on 10 µg of HeLa cell lysate.

Target description

~56 kDa observed. The calculated molecular weight is 43 kDa, however TFG has been shown as a ~56 kDa band in western blots (Greco, A., et al. (1995). Molecular and Cellular Biology. 15(11):6118–6127.)

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service