Skip to Content
Merck
All Photos(2)

Key Documents

112252

Sigma-Aldrich

Acidic Mammalian Chitinase Inhibitor, Bisdionin F

The Acidic Mammalian Chitinase Inhibitor, Bisdionin F controls the biological activity of Acidic Mammalian Chitinase.

Synonym(s):

Acidic Mammalian Chitinase Inhibitor, Bisdionin F, AMCase Inhibitor, 3,7-dimethyl-1-(3-(3-methyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)propyl)-1H-purine-2,6(3H,7H)-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 245.00

CHF 245.00


Estimated to ship on28 May 2025



Select a Size

Change View
5 MG
CHF 245.00

About This Item

Empirical Formula (Hill Notation):
C16H18N8O4
CAS Number:
Molecular Weight:
386.37
UNSPSC Code:
12352106
NACRES:
NA.54

CHF 245.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL, clear, nearly colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

CN(C=N1)C2=C1N(C)C(N(C2=O)CCCN3C(N(C)C(N=CN4)=C4C3=O)=O)=O

InChI key

PUHJHZQYIBJCSQ-UHFFFAOYSA-N

General description

A cell-permeable, competitive Acidic Mammalian Chitinase (AMCase) Inhibitor (IC50 = 0.92 µM and Ki = 0.42 µM for hAMCase, and IC50= 2.2 µM for mAMCase, in vitro) that demonstrates 20-fold selectivity for hAMCase over hCHIT1. It is shown to decrease chitinase enzymatic activity (5 mg/kg, i.p.) in the lungs of control PBS- and OVA-challenged mice. Furthermore, it attenuates lung chitinase activity, reduces eosinophil influx, and improves ventilatory function, in vivo, in a murine model of allergic inflammation. It also causes neutrophilia in the lungs of OVA-challenged mice.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Sutherland, T.E., et al. 2011. Chem. Biol.18, 569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifan Wu et al.
PLoS neglected tropical diseases, 18(2), e0011930-e0011930 (2024-02-07)
Ascariasis (roundworm) is the most common parasitic helminth infection globally and can lead to significant morbidity in children including chronic lung disease. Children become infected with Ascaris spp. via oral ingestion of eggs. It has long been assumed that Ascaris

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service