Skip to Content
Merck
All Photos(1)

Key Documents

810791

Sigma-Aldrich

Probumin® Bovine Serum Albumin Polymer Enhanced, Low Sodium Chloride, 30% solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41141801
eCl@ss:
32011401

biological source

bovine serum

Quality Level

type

for cell culture

shelf life

36 mo.

storage temp.

2-8°C

General description

A 30% solution of moderately polymerized purified BSA manufactured by a proprietary heat-shock fractionation process. It is solvent treated, extensively deionized to reduce calcium, and is concentrated. No artificial avidity enhancers or high molecular weight agglutination potentiators are added. Contains sodium azide.

Storage and Stability

refrigerated

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin BSA grades are distinguished by differences in production. The use of terms such as ‘Diagnostic Grade′ or ‘Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin BSA grades using the specific reagents, conditions and constraints of their assay.For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service