Skip to Content
Merck
All Photos(1)

Documents

81007N

Sigma-Aldrich

Probumin® Bovine Serum Albumin Microbiological Grade, 30% Solution

29-31 g/dL protein (Biuret), USA origin, microbiological culture, cell culture | mammalian: suitable, enzyme immunoassay: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801
NACRES:
NA.25

product name

Probumin® Bovine Serum Albumin Microbiological Grade, 30% Solution, pack of 50 mL

description

Does Not Contain Preservative

Quality Level

Assay

29-31 g/dL protein basis (biuret)

form

solution

shelf life

36 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pack of 50 mL

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL protein (Biuret)

technique(s)

cell culture | mammalian: suitable
enzyme immunoassay: suitable
microbiological culture: suitable
western blot: suitable

pH

6.8-7.4

foreign activity

Microbial Content Tested by USP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service