Skip to Content
Merck
All Photos(1)

Key Documents

567707

Sigma-Aldrich

Sphingomyelinase, Staphylococcus aureus

Synonym(s):

Sphingomyelinase, Staphylococcus aureus

Sign Into View Organizational & Contract Pricing

Select a Size

10 U
CHF 233.00

CHF 233.00


Estimated to ship on26 May 2025



Select a Size

Change View
10 U
CHF 233.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

CHF 233.00


Estimated to ship on26 May 2025


biological source

Staphylococcus aureus

Quality Level

form

liquid

specific activity

≥100 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Native sphingomyelinase from Staphylococcus species. May be applied exogenously to initiate the sphingomyelin signaling pathway. Has very little activity on phosphatidylcholine.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of trinitrophenylaminolauryl (TNPAL)-sphingomyelin per min at 37°C, pH 7.4.

Physical form

In 250 mM phosphate buffer, 50% glycerol, pH 7.5.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Galve-Roperh, I., et al. 1997. Brain Res. Mol. Brain Res.52, 90.
Jarvis, W.D., et al. 1994. Proc. Natl. Acad. Sci. USA 91, 73.
Mathias, S., et al. 1993. Science259, 519.
Raines, M.A., et al. 1993. J. Biol. Chem.268, 14572.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service