Skip to Content
Merck
All Photos(1)

Key Documents

1.15262

Supelco

Amberlite IRA-410 Ion Exchange Resin

15-50 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

Ion exchanger Amberlite IRA-410, suitable for ion chromatography

form

resin

Quality Level

technique(s)

HPLC: suitable
LPLC: suitable
ion chromatography: suitable (strongly basic)

matrix

PS-DVB

matrix active group

SAX

particle size

300-1100 μm

separation technique

anion exchange

storage temp.

no temp limit

General description

(strongly basic anion exchanger type II) Cl⁻ form

Analysis Note

Description: damp brownish-yellow spherules
Total capacity: ≥ 1.3 mol/l
Particle size (0,3-1,1 mm): ≥ 90

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service