1.04765
I Ion Exchange Resin
hydrogen form, 15-50 mesh, strongly acidic, cationic
Synonym(s):
polymeric adsorbent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Ion exchanger I, (strongly acidic cation exchanger, H⁺-form) GR for analysis Reag. Ph Eur
Agency
reag. Ph. Eur.
Quality Level
form
resin
technique(s)
HPLC: suitable
LPLC: suitable
matrix
PS-DVB
matrix active group
SCX
particle size
300-1100 μm
bulk density
850 kg/m3
separation technique
cation exchange
storage temp.
no temp limit
Related Categories
Analysis Note
Appearance: yellow brown, transparent spherules
Exchange capacity: ≥ 1.7 mol/l
Exchange capacity (calculated on dried substance): 4.5 - 5
Water (according to Karl Fischer): 50 - 60
Exchange capacity: ≥ 1.7 mol/l
Exchange capacity (calculated on dried substance): 4.5 - 5
Water (according to Karl Fischer): 50 - 60
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service