Skip to Content
Merck
All Photos(1)

Key Documents

09-230

Sigma-Aldrich

Anti-phospho-Raptor (Thr706) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

P150 target of rapamycin (TOR)-scaffold protein, p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, regulatory associated protein of mTOR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

canine, human

species reactivity (predicted by homology)

horse (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), dog (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr706)

Gene Information

bovine ... Rptor(507056)
human ... RPTOR(57521)

General description

Raptor is a subunit of mTORC1 which regulates cell growth and survival in response to nutrient and hormonal signals. mTORC1 is activated in response to growth factors or amino-acids. Within mTORC1, Raptor positively regulates mTORC1 activity and functions as a scaffold for recruiting mTORC1 substrates.

Specificity

This antibody recognizes Raptor phosphorylated at Thr706.

Immunogen

Epitope: Ser706
KLH-conjugated linear peptide corresponding to human Raptor phosphorylated at and around Thr706.

Application

Detect phospho-Raptor (Thr706) using this Anti-phospho-Raptor (Thr706) Antibody validated for use in WB, DB.

Quality

Evaluated by Dot Blot in modified and non modified peptides of phosphorylated and non-phosphorlyated Raptor protein.

Dot Blot Analysis: 1 µg/mL of this antibody detected Raptor in modified and non-modified peptides of phosphorylated and non-phosphorlyated Raptor protein.

Target description

~ 150 kDa

Physical form

Purified rabbit serum in PBS with 0.05% sodium azide.

Analysis Note

Control
Modified and non modified peptides of phosphorylated and non-phosphorlyated Raptor protein

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shijie Ma et al.
Oncogene, 41(17), 2431-2443 (2022-03-14)
eIF3a (eukaryotic translation initiation factor 3a), a subunit of the eIF3 complex, has been suggested to play a regulatory role in protein synthesis and in cellular response to DNA-damaging treatments. S6K1 is an effector and a mediator of mTOR complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service