Skip to Content
Merck
All Photos(2)

Key Documents

860394P

Avanti

18:0-18:0-d35 PC

Avanti Research - A Croda Brand 860394P, powder

Synonym(s):

1-stearoyl-2-stearoyl-d35-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 999.00

CHF 999.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CHF 999.00

About This Item

Empirical Formula (Hill Notation):
C44H53NO8PD35
CAS Number:
Molecular Weight:
825.36
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

CHF 999.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860394P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860394P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.
Phosphocholine (PC) is a key component and an end product of phosphatidylcholine (lecithin) degradation, which is a main phospholipid of eukaryotic bio-membranes.[1]

Biochem/physiol Actions

Phosphocholine (PC) and PC modified lipids stimulate complement fixation and other effector reactions implicated in host defense by interacting with C-reactive protein (CRP) or natural antibodies.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860394P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphocholine-an agonist of metabotropic but not of ionotropic functions of alpha 9-containing nicotinic acetylcholine receptors
Richter K, et al.
Scientific reports, 6, 28660-28660 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service