Skip to Content
Merck
All Photos(2)

Key Documents

860366P

Avanti

18:0 PC-d74

Avanti Research - A Croda Brand, powder

Synonym(s):

1,2-Distearoyl-d70-sn-glycero-3-phosphocholine-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 1’060.00

CHF 1’060.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CHF 1’060.00

About This Item

Empirical Formula (Hill Notation):
C44H14NO8PD74
CAS Number:
Molecular Weight:
864.60
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

CHF 1’060.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860366P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C)(C)C)=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(

General description

18:0 PC-d74 is a deuterated compound of phosphorylcholine. Phosphorylcholine is present in several microbes. It is a small zwitter ionic amino alcohol.

Application

18:0 PC-d74 has been used as a component of the lipid bilayer for sum frequency generation vibrational spectroscopy (SFG-VS) studies.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860366P-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Observing Peptide-Induced Lipid Accumulation in a Single-Component Zwitterionic Lipid Bilayer
Streptococcus Pneumoniae: molecular mechanisms of host-pathogen interactions, 119(51), 28523-28529 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service