Skip to Content
Merck
All Photos(2)

Documents

856705P

Avanti

16:0 Lyso PE

Avanti Research - A Croda Brand, powder

Synonym(s):

1-hexadecanoyl-sn-glycero-3-phosphoethanolamine; PE(16:0/0:0); 110698, Lysophosphatidylethanolamine 16:0

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H44NO7P
CAS Number:
Molecular Weight:
453.55
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (856705P-200mg)
pkg of 1 × 25 mg (856705P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCCCCCCCCCC)=O

Application

16:0 Lyso PE has been used to degranulate human eosinophils. It has also been used to enhance nerve growth factor (NGF)-induced mitogen-activated protein kinase (MAPK) phosphorylation.

Packaging

5 mL Amber Glass Screw Cap Vial (856705P-200mg)
5 mL Amber Glass Screw Cap Vial (856705P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine enhances NGF-induced MAPK and Akt signals through the extracellular domain of TrkA in PC12 cells
Itakura A, et al.
FEBS Open Bio, 3(1), 243-251 (2013)
Miki Eto et al.
International journal of molecular sciences, 13(12), 16267-16280 (2012-12-05)
Cellular membranes contain glycerophospholipids, which have important structural and functional roles in cells. Glycerophospholipids are first formed in the de novo pathway (Kennedy pathway) and are matured in the remodeling pathway (Lands' cycle). Recently, lysophospholipid acyltransferases functioning in Lands' cycle
Yoko Shima et al.
The Journal of biological chemistry, 295(20), 6983-6991 (2020-04-10)
Newly synthesized major histocompatibility complex (MHC) class I proteins are stabilized in the endoplasmic reticulum (ER) by binding 8-10-mer-long self-peptide antigens that are provided by transporter associated with antigen processing (TAP). These MHC class I:peptide complexes then exit the ER
Qingxin Shi et al.
Food & function, 10(12), 7782-7792 (2019-11-30)
Triterpenoid saponins from Kuding tea have demonstrated preventive effects on hyperlipidaemia induced by a high-fat diet. Lysoglycerophospholipids (Lyso-GPLs) are known to be associated with proatherogenic conditions such as hyperlipidaemia. In this study, a target profiling strategy based on a multiple
Wuhanqimuge et al.
FEBS open bio, 3, 243-251 (2013-06-19)
Lysophosphatidylcholine (LPC) is one of the major lysophospholipids mainly generated by phospholipase A2 (PLA2)-mediated hydrolysis of phosphatidylcholine (PC). We previously found that LPC displays neurotrophin-like activity in the rat pheochromocytoma PC12 cells and in cerebellar granule neurons, but the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service