Merck
  • Lysophosphatidylcholine acyltransferase 3 is the key enzyme for incorporating arachidonic acid into glycerophospholipids during adipocyte differentiation.
Lysophosphatidylcholine acyltransferase 3 is the key enzyme for incorporating arachidonic acid into glycerophospholipids during adipocyte differentiation.
International journal of molecular sciences (2012-12-05)
Miki Eto, Hideo Shindou, Andreas Koeberle, Takeshi Harayama, Keisuke Yanagida, Takao Shimizu
ABSTRACT
Cellular membranes contain glycerophospholipids, which have important structural and functional roles in cells. Glycerophospholipids are first formed in the de novo pathway (Kennedy pathway) and are matured in the remodeling pathway (Lands' cycle). Recently, lysophospholipid acyltransferases functioning in Lands' cycle were identified and characterized. Several enzymes involved in glycerophospholipid biosynthesis have been reported to have important roles in adipocytes. However, the role of Lands' cycle in adipogenesis has not yet been reported. Using C3H10T1/2, a cell line capable of differentiating to adipocyte-like cells in vitro, changes of lysophospholipid acyltransferase activities were investigated. Lysophosphatidylcholine acyltransferase (LPCAT), lysophosphatidylethanolamine acyltransferase (LPEAT) and lysophosphatidylserine acyltransferase (LPSAT) activities were enhanced, especially with 18:2-CoA and 20:4-CoA as donors. Correspondingly, mRNA expression of LPCAT3, which possesses LPCAT, LPEAT and LPSAT activities with high specificity for 18:2- and 20:4-CoA, was upregulated during adipogenesis. Analysis of acyl-chain compositions of phosphatidylcholine (PC), phosphatidylethanolamine (PE) and phosphatidylserine (PS) showed a change in their profiles between preadipocytes and adipocytes, including an increase in the percentage of arachidonic acid-containing phospholipids. These changes are consistent with the activities of LPCAT3. Therefore, it is possible that enhanced phospholipid remodeling by LPCAT3 may be associated with adipocyte differentiation.
Product Description
Avanti
16:0 Lyso PS, 1-palmitoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), powder
Avanti
16:0 Lyso PE, Avanti Research - A Croda Brand, powder
Avanti
17:0-20:4 PS, Avanti Research - A Croda Brand LM1302, methanol solution