Skip to Content
Merck
All Photos(2)

Key Documents

700036P

Avanti

4β-hydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3b,4b-diol; 110803

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 127.00
10 MG
CHF 226.00

CHF 127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 127.00
10 MG
CHF 226.00

About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 127.00


Check Cart for Availability

Request a Bulk Order

description

cholest-5-ene-3β,4β-diol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (700036P-10mg)
pkg of 1 × 5 mg (700036P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC1CCC2(C3C(C4C(C(CC4)C(CCCC(C)C)C)(CC3)C)CC=C2C1O)C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)20-11-12-21-19-9-10-23-25(29)24(28)14-16-27(23,5)22(19)13-15-26(20,21)4/h10,17-22,24-25,28-29H,6-9,11-16H2,1-5H3

InChI key

CZDKQKOAHAICSF-UHFFFAOYSA-N

General description

4β-hydroxycholesterol is an endogenous oxysterol, which is formed by cytochrome P450 3A (CYP3A4) and CYP3A5.[1]

Biochem/physiol Actions

4β-hydroxycholesterol acts as a marker for cytochrome P450 3A (CYP3A4/5) activity.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700036P-10mg)
5 mL Amber Glass Screw Cap Vial (700036P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4beta-hydroxycholesterol as an endogenous marker for CYP3A4/5 activity. Stability and half-life of elimination after induction with rifampicin
Diczfalusy U, et al.
British Journal of Clinical Pharmacology, 67(1), 38-43 (2009)
Michael E Abrams et al.
Nature microbiology, 5(7), 929-942 (2020-04-15)
Cholesterol 25-hydroxylase (CH25H) is an interferon-stimulated gene that converts cholesterol to the oxysterol 25-hydroxycholesterol (25HC). Circulating 25HC modulates essential immunological processes including antiviral immunity, inflammasome activation and antibody class switching; and dysregulation of CH25H may contribute to chronic inflammatory disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service