Key Documents
Z303186
Quickfit® Liebig condenser
L 250 mm, joint: ST/NS 19/26
Synonym(s):
C1/12/25/SC
Select a Size
1 EA
CHF 59.50
CHF 59.50
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 59.50
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
CHF 59.50
Check Cart for Availability
Joint
joint: ST/NS19/26
manufacturer/tradename
Quickfit C1/12/25/SC
availability
not available in USA and Canada
not available in USA and Canada
L
250 mm
surface area
130 cm2
General description
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)