M000037A0
Empty 37 mm Monitor Cassette with rings and plugs
Reload cassette with appropriate filters
Synonym(s):
Empty cassette
About This Item
Recommended Products
material
polystyrene
Quality Level
sterility
non-sterile
filter diam.
37 mm
filtration area
9 cm2
fitting
female Luer inlet slip
female Luer outlet slip
application(s)
environmental
shipped in
ambient
General description
•Empty aerosol analysis monitors
•For use with Type MF (mixed cellulose esters) Millipore filters
Millipore′s 37 mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders for use with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface.
A three-part design holds the filter in-place when the top section is removed for open aerosol sampling. The Average Background particle Count (ABC) value is marked on each monitor package.
Applications:
Aerosol analysis monitors are available with 0.45 µm and 0.8 µm cellulose esters membranes. Matched weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test filter pre-weighing in gravimetric analysis.
Application
Principle
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service