Documents
WHA10451710
Whatman® pressure filtration devices
MD 142/7/3 PTFE filter holder, 1500 mL infusion vessel, capacity 142 mm
Synonym(s):
Z751294
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
PTFE housing
feature
1500 mL infusion vessel
packaging
pk of 1
manufacturer/tradename
Whatman 10451710
Whatman Article No. 28415695 (US reference)
capacity
142 mm
General description
Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders are connected inline and are suitable for larger volumes of several liters.
Filtration of liquids and gases is possible, including sterile filtration* of serums or clear filtration of media that are difficult to filter, especially those that are highly viscous.
Features and Benefits
  • Membranes, paper or glass fiber filter discs can be used.
  • Cleaning and changing of filters is completed in a few steps.
Other Notes
Field of Use: For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva