Skip to Content
Merck
All Photos(2)

Key Documents

P45702

Sigma-Aldrich

(−)-α-Pinene

98%, optical purity ee: ≥81% (GLC)

Synonym(s):

(-)-alpha-Pinene, (1S)-(−)-α-Pinene, (1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1S,5S)-2-Pinene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 116.00
500 ML
CHF 208.00

CHF 116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CHF 116.00
500 ML
CHF 208.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1903790
EC Number:
MDL number:
UNSPSC Code:
12352002
eCl@ss:
39011007
PubChem Substance ID:
NACRES:
NA.22

CHF 116.00


Check Cart for Availability

Request a Bulk Order

vapor density

4.7 (vs air)

Quality Level

vapor pressure

~3 mmHg ( 20 °C)

Assay

98%

optical activity

[α]22/D −42°, neat

optical purity

ee: ≥81% (GLC)

autoignition temp.

491 °F

refractive index

n20/D 1.465 (lit.)

bp

155-156 °C (lit.)

mp

-64 °C (lit.)

density

0.874 g/mL at 25 °C

SMILES string

CC1=CCC2CC1C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m0/s1

InChI key

GRWFGVWFFZKLTI-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

88.3 °F

Flash Point(C)

31.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service