Skip to Content
Merck
(−)-α-Pinene 98%, optical purity ee: ≥81% (GLC)

(−)-α-Pinene

(−)-α-Pinene 98%, optical purity ee: ≥81% (GLC)
Synonym(s):
(1S)-(−)-α-Pinene, (1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1S,5S)-2-Pinene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique