Skip to Content
Merck
All Photos(2)

Key Documents

MT30207953

Mettler Toledo Seven2Go portable pH meter S2

Light Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 710.00

CHF 710.00


Check Cart for Availability


Select a Size

Change View
1 KIT
CHF 710.00

About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

CHF 710.00


Check Cart for Availability

description

connector BNC/RCA
pH/ion/ORP meters

feature

Light Kit

manufacturer/tradename

Mettler-Toledo 30207953

ORP measuring range

-1999-1999 mV

pH measuring range

-2-20

temperature measuring range

-5-105 °C

battery power

4 x 1.5 V AA (or 1.3 V NiMH accumulators)

Looking for similar products? Visit Product Comparison Guide

General description

Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.

Simple Menu – Intuitive Operation:
  • Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions

T-Pad – Comfortable, Fast Navigation:
  • Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement

ISM® Ready – Reduced Chances for Error:
  • Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data

IP67 – Weatherproof and Robust Design:
  • Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
  • 5-point pH callibration

Application

Students and educational purpose

Features and Benefits

Includes: Meter, electrode clip, instructions, quick guide, test certificate, declaration of conformity, batteries, InLab Versatile Pro electrode, bottles for callibration, buffer sachets for pH 4.01, 7.00 and 9.21 and uGo carry case

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service