MT30014092
Mettler-Toledo InLab® 73 series conductivity probe
InLab® 731-ISM, With ISM, cable L 1 m, Mini-DIN connector, IP67
Select a Size
CHF 730.00
Select a Size
About This Item
CHF 730.00
Recommended Products
material
epoxy shaft
description
4 pole graphite cell
cell constant 0.57 cm-1, epoxy shaft
feature
Mini-DIN connector, IP67
With ISM
manufacturer/tradename
Mettler-Toledo 30014092
L × diam.
120 mm × 12 mm
cable L
1 m
Looking for similar products? Visit Product Comparison Guide
General description
Supplied with quality certificate, integrated temperature sensor, plug and cable.
Intended use and principles
These products combine a state-of-the-art conductivity cell with a thermistor which is integrated in the sensor body enabling automatic temperature compensating (ATC) to provide an easy to use, reliable and accurate sensor. The electrode design minimizes the potential for external interferences from sample containers or other sensors. These
sensors are equipped with cables in various lengths and two types of plugs to match Mettler Toledo conductivity meters, with and without Intelligent Sensor Management (ISM®).
Intelligent Sensor Management (ISM)
The Mettler Toledo dual channel portable meters SevenGo Duo (pro) SG23, 68 and 78 have the ability to automatically recognize ISM sensors upon connection, transfer calibration data to and from the sensor etc. For more information regarding ISM please read the respective chapter of the operating instructions supplied with the meter.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service