Key Documents
MT30014098
Mettler Toledo flow-cell for InLab® trace conductivity probe
Select a Size
1 EA
CHF 1’750.00
CHF 1’750.00
Estimated to ship on09 April 2025
Select a Size
Change View
1 EA
CHF 1’750.00
About This Item
UNSPSC Code:
41115600
NACRES:
NB.79
CHF 1’750.00
Estimated to ship on09 April 2025
manufacturer/tradename
Mettler-Toledo 30014098
General description
Using the Flow Cell the InLab® Trace the highest repeatability and reproducibility ever can be achieved.
The Flow Cell InLab® Trace was built at an angle to prevent air bubbles being trapped. Your workflow is optimized to the max. Enjoy the peace of mind.
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)