Skip to Content
Merck
All Photos(1)

Key Documents

F505

Sigma-Aldrich

Fisetin

Synonym(s):

3,3′,4′,7-Tetra­hydroxy­flavone, 5-Deoxyquercetin, Natural Brown 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6 · xH2O
CAS Number:
Molecular Weight:
286.24 (anhydrous basis)
Beilstein:
292829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

>330 °C (lit.)

SMILES string

O.Oc1ccc2C(=O)C(O)=C(Oc2c1)c3ccc(O)c(O)c3

InChI

1S/C15H10O6.H2O/c16-8-2-3-9-12(6-8)21-15(14(20)13(9)19)7-1-4-10(17)11(18)5-7;/h1-6,16-18,20H;1H2

InChI key

GYHFUROKCOMWNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Basic & clinical pharmacology & toxicology, 115(4), 321-329 (2014-03-22)
Intravesical chemotherapy after transurethral resection has been widely used as an adjuvant therapy to prevent recurrence and progression of superficial bladder cancer. Due to the insufficiency of the current chemotherapeutics, there is an urgent need to search for more novel
Yewseok Suh et al.
Carcinogenesis, 30(2), 300-307 (2008-11-28)
Overexpression of cyclooxygenase 2 (COX2) and uncontrolled wingless and Int (Wnt)-signaling pathway have long been suggested to play crucial roles in colorectal cancer. Studies show that selective COX2 inhibitors possess great potential as chemopreventive agents for colon cancer. Recent studies
Ragini Sinha et al.
European journal of medicinal chemistry, 79, 102-109 (2014-04-15)
Flavonols are an important class of naturally occurring molecules and are known for their pharmacological activity. The activity is associated with the ability of flavonols to influence membrane-dependent processes. We have investigated the in-vitro anti-proliferative and anti-oxidant activity of galangin
Stephanie Lemmo Ham et al.
Integrative biology : quantitative biosciences from nano to macro, 7(7), 792-800 (2015-06-30)
Cell migration is a major process that drives metastatic progression of cancers, the major cause of cancer death. Existing chemotherapeutic drugs have limited efficacy to prevent and/or treat metastasis, emphasizing the need for new treatments. We focus on triple negative
Niranjan Koirala et al.
Journal of biotechnology, 184, 128-137 (2014-05-27)
An O-methyltransferase SpOMT2884, originating from Streptomyces peucetius ATCC 27952, was cloned, expressed, and applied for the production of target metabolite from Escherichia coli. Biochemical characterization of the 25kDa recombinant protein by in vitro and in vivo experiments showed that SpOMT2884

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service