MilliporeSigma
Methylation and subsequent glycosylation of 7,8-dihydroxyflavone.
Journal of biotechnology (2014-05-27)
Niranjan Koirala, Ramesh Prasad Pandey, Prakash Parajuli, Hye Jin Jung, Jae Kyung Sohng
ABSTRACT
An O-methyltransferase SpOMT2884, originating from Streptomyces peucetius ATCC 27952, was cloned, expressed, and applied for the production of target metabolite from Escherichia coli. Biochemical characterization of the 25kDa recombinant protein by in vitro and in vivo experiments showed that SpOMT2884 was an S-adenosyl-l-methionine-dependent O-methyltransferase. SpOMT2884 catalyzed O-methylation of different classes of flavonoids such as flavones (7,8-dihydroxyflavone (7,8-DHF), luteolin), flavonols (quercetin, rutin), flavanone (naringenin), and isoflavonoids (daidzein, formononetin). Biotransformation of 7,8-DHF, a preferred substrate of SpOMT2884, in a grown-induced culture of E. coli BL21 (DE3) harboring the recombinant pET-28a-SpOMT2884 stoichiometrically converted 7,8-DHF into 7-hydroxy-8-methoxyflavone, which was confirmed by liquid chromatography, mass spectrometry and various nuclear magnetic resonance (NMR) spectroscopy analyses. In order to improve the biotransformation substrate, time and media parameters were optimized and the production was scaled up using a 3-L fermentor. The maximum yield of 7-hydroxy-8-methoxyflavone was 192μM (52.57mg/L), representing almost 96% bioconversion within 12h, when 200μM of 7,8-DHF was supplemented in the culture. Further, the 7-hydroxy-8-methoxyflavone was purified in large scale and was used as a substrate separately for in vitro glycosylation to produce glucose, galactose and 2-deoxyglucose conjugated at 7th hydroxyl position of 7-hydroxy-8-methoxyflavone. Biological activity showed that 7-hydroxy-8-methoxyflavone had long term cytoprotective and antioxidant effects compared to 7,8-DHF suggesting that methylation enhances the stability of substrate and glycosylation has proved to increase the water solubility.
Product Description
Supelco
Daidzein, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Luteolin, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Daidzein, ≥98%, synthetic
SKU
Pack Size
Availability
Price
Quantity
Luteolin, primary reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid