Skip to Content
Merck
All Photos(1)

Documents

933422

Sigma-Aldrich

AFM Disc

disc diam. 10 mm

Synonym(s):

Metal Discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.23

disc diam.

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

Each metal disc pack contains 50 units of metal discs.

Application

Metal Specimen Discs are used with the Atomic Force Microscope. These are high quality metal discs with smooth edges and flat surfaces for use in atomic force microscopy.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service