Skip to Content
Merck
All Photos(2)

Key Documents

911828

Sigma-Aldrich

PhPAd-DalPhos Ni(o-tolyl)Cl

≥95%

Synonym(s):

(SP-4-3)-Chloro[8-[2-(diphenylphosphino-κP)phenyl]-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.13,7]decane-κP8](2-methylphenyl)nickel

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 632.00

CHF 632.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
CHF 632.00

About This Item

Empirical Formula (Hill Notation):
C35H37ClNiO3P2
CAS Number:
Molecular Weight:
661.76
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

CHF 632.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

235-248 °C

SMILES string

Cl[Ni]1(P(C(C2)(O3)C)(C4=CC=CC=C4P(C5=CC=CC=C5)1C6=CC=CC=C6)[C@@](C[C@]3(OC27C)C)(O7)C)C(C=CC=C8)=C8C

Application

PhPAd-DalPhos Ni(o-tolyl)Cl is an air stable precatalyst developed in the Stradiotto lab used for coupling of bulky primary amines with arenes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2 - Repr. 2 - Resp. Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph P Tassone et al.
Angewandte Chemie (International ed. in English), 58(8), 2485-2489 (2019-01-04)
The base metal-catalyzed C-N cross-coupling of bulky α,α,α-trisubstituted primary alkylamines with (hetero)aryl electrophiles represents a challenging and under-developed class of transformations that is of significant potential utility, including in the synthesis of lipophilic active pharmaceutical ingredients. Herein, we report that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service