Skip to Content
Merck
All Photos(2)

Key Documents

796565

Sigma-Aldrich

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid

95%

Synonym(s):

Fenofibric acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 45.20

CHF 45.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CHF 45.20

About This Item

Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75
Beilstein:
2058973
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 45.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

mp

187 °C

functional group

carboxylic acid
chloro
ketone

SMILES string

O=C(C1=CC=C(OC(C)(C)C(O)=O)C=C1)C2=CC=C(Cl)C=C2

InChI

1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)

InChI key

MQOBSOSZFYZQOK-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasser Majeed et al.
Scientific reports, 9(1), 6316-6316 (2019-04-21)
Angiosarcomas are highly aggressive tumors of endothelial origin, which carry a poor prognosis. Fenofibrate is a hypolipidemic drug, which acts by activating the transcription factor PPARα. It has also been widely reported to have 'anti-cancer' activity. The current study investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service